Gene Transcripts

Gene Symbol
Sec22a
Entrez Gene ID
317717
Gene Name
SEC22 vesicle trafficking protein homolog A (S. cerevisiae)
Gene Aliases
1810005C06Rik, Sec22l2
Chromosome Location
Chr. 16: 35311131 - 35363934 on Build GRCm38
UniGene ID
Mm.259164
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133704.4 NP_598465.2 2 245
GenBank mRNA AK007341.1 - 2 185
AK037540.1 - 2 180
AK044615.1 - 2 180
AK082516.1 - 2 245
BC057176.1 - 1 158
BC138969.1 - 1 159 8861991
BC138971.1 - 1 159 8861993
BC145477.1 - 1 161
BC145478.1 - 1 161 9054040

View Full Product Details