Gene Transcripts

Gene Symbol
Errfi1
Entrez Gene ID
74155
Gene Name
ERBB receptor feedback inhibitor 1
Gene Aliases
1300002F13Rik, AI788755, Mig-6, Mig6, RALT
Chromosome Location
Chr. 4: 150853801 - 150868889 on Build GRCm38
UniGene ID
Mm.318841
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133753.1 NP_598514.1 4 583
GenBank mRNA AK004851.1 - 4 602
AK139780.1 - 4 599
AK166791.1 - 4 602
AK167156.1 - 4 542
AK167359.1 - 4 602
BC005546.1 - 4 583 3585640
BC057646.1 - 4 601 4194124

View Full Product Details