Gene Transcripts

Gene Symbol
Fbxl14
Entrez Gene ID
101358
Gene Name
F-box and leucine-rich repeat protein 14
Gene Aliases
AW322056, Fbx14l
Chromosome Location
Chr. 6: 119479668 - 119483886 on Build GRCm38
UniGene ID
Mm.125477
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133940.3 NP_598701.2 1 2568
GenBank mRNA AK036124.1 - 1 2465
AK084506.1 - 1 2455
AK155154.1 - 1 2440
BC006913.1 - 1 2145
BC021329.1 - 1 2449 3710945

View Full Product Details