Gene Transcripts

Gene Symbol
Fam50a
Entrez Gene ID
108160
Gene Name
family with sequence similarity 50, member A
Gene Aliases
D0HXS9928E, XAP-5
Chromosome Location
Chr. X: 74313033 - 74320149 on Build GRCm38
UniGene ID
Mm.4370
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138607.3 NP_613073.2 1, 2 301
GenBank mRNA AK008402.1 - 1, 2 302
BC026128.1 - 1, 2 211
Y18505.1 - 1, 2 266

View Full Product Details