Gene Transcripts

Gene Symbol
Rhbdl3
Entrez Gene ID
246104
Gene Name
rhomboid, veinlet-like 3 (Drosophila)
Gene Aliases
AI847581, Rhbdl4, Rhomr1, Vrho
Chromosome Location
Chr. 11: 80300432 - 80355986 on Build GRCm38
UniGene ID
Mm.194148
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139228.3 NP_631974.1 4, 5 776
GenBank mRNA AJ313479.1 - 4, 5 776
AK041678.1 - 2, 3 415
BC054784.1 - 4, 5 612 6817213
BC056969.1 - 4, 5 612 6817213

View Full Product Details