Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139236.3 | NP_631982.2 | 2 | 228 | |
GenBank mRNA | AF361077.1 | - | 2 | 177 | |
AF361078.1 | - | 2 | 177 | ||
AK053117.1 | - | 2 | 345 | ||
AK089138.1 | - | 2 | 148 | ||
AK155818.1 | - | 2 | 167 | ||
AK166559.1 | - | 2 | 154 | ||
AK166650.1 | - | 2 | 154 | ||
AK171163.1 | - | 2 | 165 | ||
AK171421.1 | - | 2 | 155 | ||
AK191861.1 | - | 2 | 84 | ||
AK195706.1 | - | 2 | 84 | ||
AK197974.1 | - | 2 | 84 | ||
AK198310.1 | - | 2 | 84 | ||
AK199838.1 | - | 2 | 84 | ||
AK202668.1 | - | 2 | 84 | ||
AK209500.1 | - | 2 | 84 | ||
BC059820.1 | - | 2 | 146 | 6827596 | |
BC060189.1 | - | 2 | 156 | ||
CN459855.1 | - | 2 | 228 |