Gene Transcripts

Gene Symbol
Hnrnpll
Entrez Gene ID
72692
Gene Name
heterogeneous nuclear ribonucleoprotein L-like
Gene Aliases
2510028H02Rik, 2810036L13Rik, AI256697, AI852082, Hnrpll
Chromosome Location
Chr. 17: 80029487 - 80062513 on Build GRCm38
UniGene ID
Mm.64579
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144802.4 NP_659051.3 10, 11 1833
GenBank mRNA AK012866.1 - 10, 11 1549
AK146879.1 - 10, 11 1795
BC004763.1 - 7, 8 806
BC006805.1 - 1, 2 1505
BC012849.1 - 10, 11 1779 3587716
BC038912.1 - 2, 3 325

View Full Product Details