Gene Transcripts

Gene Symbol
ADH1B
Entrez Gene ID
125
Gene Name
alcohol dehydrogenase 1B (class I), beta polypeptide
Gene Aliases
ADH2, HEL-S-117
Chromosome Location
Chr. 4: 99306387 - 99321442 on Build GRCh38
UniGene ID
Hs.4
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000668.5 NP_000659.2 8 1101
NM_001286650.1 NP_001273579.1 9 1223
GenBank mRNA AF153821.1 - 8 1058
AK299409.1 - 9 1019
AK301018.1 - 9 1182
AK303945.1 - 7 891
AK304396.1 - 8 825
AK316501.1 - 8 891
BC033009.2 - 8 1060 4828235
BX647469.1 - 8 3085
D00137.1 - 8 1010
FJ224285.1 - 8 1060
M21692.1 - 8 1022
M24317.1 - 8 1020
X03350.1 - 8 1062

View Full Product Details