Gene Transcripts

Gene Symbol
Crtac1
Entrez Gene ID
72832
Gene Name
cartilage acidic protein 1
Gene Aliases
2810454P21Rik, AW047536, Crtac1B, Lotus
Chromosome Location
Chr. 19: 42283036 - 42431820 on Build GRCm38
UniGene ID
Mm.313558
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145123.4 NP_660105.3 3 545
GenBank mRNA AJ421516.1 - 3 395
AK032328.1 - 3 410
AK049801.1 - 3 516
AK083221.1 - 3 548
AK149020.1 - 3 551
AK149069.1 - 3 546

View Full Product Details