Gene Transcripts

Gene Symbol
Slc25a28
Entrez Gene ID
246696
Gene Name
solute carrier family 25, member 28
Gene Aliases
2210403D18Rik, Mfrn2, Mrs3/4
Chromosome Location
Chr. 19: 43663801 - 43677099 on Build GRCm38
UniGene ID
Mm.23827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145156.1 NP_660138.1 2 471
GenBank mRNA AF377994.1 - 2 289
AK155044.1 - 2 450
BC025908.1 - 2 471 5043992

View Full Product Details