Gene Transcripts

Gene Symbol
Tmem51
Entrez Gene ID
214359
Gene Name
transmembrane protein 51
Gene Aliases
BC003277
Chromosome Location
Chr. 4: 142030990 - 142084954 on Build GRCm38
UniGene ID
Mm.27587
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145402.3 NP_663377.1 3 1191
GenBank mRNA AK077758.1 - 3 1191
AK164026.1 - 3 1133
BC003277.1 - 3 1055 3582174

View Full Product Details