Gene Transcripts
- Gene Symbol
- Cyp2j13
- Entrez Gene ID
- 230459
- Gene Name
- cytochrome P450, family 2, subfamily j, polypeptide 13
- Gene Aliases
- Chromosome Location
- Chr. 4: 96042660 - 96077566 on Build GRCm38
- UniGene ID
- Mm.268175
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC016446.1 |
- |
7 |
1121 |
|
View Full Product Details