Gene Transcripts

Gene Symbol
Fam76a
Entrez Gene ID
230789
Gene Name
family with sequence similarity 76, member A
Gene Aliases
Chromosome Location
Chr. 4: 132899213 - 132923226 on Build GRCm38
UniGene ID
Mm.11473
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163792.1 NP_001157264.1 8 1430
NM_145553.2 NP_663528.1 9 1517
GenBank mRNA AK037828.1 - 6 1202
AK038831.1 - 9 1525
AK154575.1 - 8 1432
AK169817.1 - 1 64
AK197668.1 - 1 114
BC008163.1 - 9 1476 3709256

View Full Product Details