Gene Transcripts

Gene Symbol
Kcnc4
Entrez Gene ID
99738
Gene Name
potassium voltage gated channel, Shaw-related subfamily, member 4
Gene Aliases
AI850292, KSHIIIC, Kcr2-4, Kv3.4
Chromosome Location
Chr. 3: 107438296 - 107459952 on Build GRCm38
UniGene ID
Mm.101976
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145922.2 NP_666034.1 2 775
GenBank mRNA AK158073.1 - 2 175
BC024837.1 - 2 775 5358950

View Full Product Details