Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195485.1 | NP_001182414.1 | 2 | 197 | |
NM_001195486.1 | NP_001182415.1 | 2 | 197 | ||
NM_001195487.1 | NP_001182416.1 | 2 | 197 | ||
NM_146083.2 | NP_666195.1 | 2 | 197 | ||
GenBank mRNA | AK034952.1 | - | 2 | 199 | |
AK045884.1 | - | 2 | 359 | ||
AK082848.1 | - | 2 | 197 | ||
AK137795.1 | - | 2 | 198 | ||
AK161652.1 | - | 2 | 196 | ||
AK168355.1 | - | 2 | 197 | ||
AK182977.1 | - | 1 | 98 | ||
BC014857.1 | - | 2 | 178 | 2646366 | |
BC024865.1 | - | 2 | 188 | ||
BC025529.1 | - | 2 | 188 | 5342587 | |
BC027391.1 | - | 2 | 185 | 4945988 | |
BY740981.1 | - | 2 | 196 |