Gene Transcripts

Gene Symbol
Tspan33
Entrez Gene ID
232670
Gene Name
tetraspanin 33
Gene Aliases
1300010A20Rik, Pen
Chromosome Location
Chr. 6: 29694214 - 29718558 on Build GRCm38
UniGene ID
Mm.225289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301407.1 NP_001288336.1 6 709
NM_146173.3 NP_666285.1 6 712
GenBank mRNA AF276890.1 - 6 711
AK004954.1 - 6 685
AK172176.1 - 6 690
BC024685.1 - 6 670 3981492
BE861472.1 - 4 321
BU054333.1 - 6 463

View Full Product Details