Gene Transcripts

Gene Symbol
Olfr31
Entrez Gene ID
18330
Gene Name
olfactory receptor 31
Gene Aliases
MOR274-1, MTPCR53
Chromosome Location
Chr. 14: 14328113 - 14329066 on Build GRCm38
UniGene ID
Mm.484338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147027.2 NP_667238.2 1 78
GenBank mRNA BC106818.1 - 1 74
BC106819.1 - 1 78 40043407
BC106824.1 - 1 78 40043472

View Full Product Details