Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152915.1 | NP_690879.1 | 2 | 981 | |
GenBank mRNA | AB067650.1 | - | 2 | 630 | |
AF370126.1 | - | 2 | 637 | ||
AK030642.1 | - | 2 | 780 | ||
AK044597.1 | - | 2 | 744 | ||
AY032924.1 | - | 2 | 637 | ||
BC034634.1 | - | 2 | 981 | 5365190 |