Gene Transcripts

Gene Symbol
Kif1c
Entrez Gene ID
16562
Gene Name
kinesin family member 1C
Gene Aliases
B430105J22Rik, D11Bwg1349e, Ltxs1
Chromosome Location
Chr. 11: 70700548 - 70731970 on Build GRCm38
UniGene ID
Mm.99996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153103.2 NP_694743.2 5 594
GenBank mRNA AB074017.1 - 3 224
AK029250.1 - 5 521
AK173007.1 - 4 301
BC029286.1 - 5 581

View Full Product Details