Gene Transcripts

Gene Symbol
Ubap2l
Entrez Gene ID
74383
Gene Name
ubiquitin-associated protein 2-like
Gene Aliases
3110083O19Rik, 4932431F02Rik, A430103N23Rik, C77168, Nice-4, mKIAA0144
Chromosome Location
Chr. 3: 89999589 - 90052623 on Build GRCm38
UniGene ID
Mm.25610
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165983.1 NP_001159455.1 4 318
NM_001165984.1 NP_001159456.1 4 293
NM_001165985.1 NP_001159457.1 4 318
NM_001165986.1 NP_001159458.1 4 293
NM_001165987.1 NP_001159459.1 4 318
NM_001165988.1 NP_001159460.1 4 293
NM_028475.2 NP_082751.1 4 318
NM_153489.2 NP_705693.2 4 318
GenBank mRNA AF276967.1 - 4 253
AK014274.1 - 4 255
AK030218.1 - 4 259
AK031934.1 - 4 298
AK040551.1 - 4 280
AK048286.1 - 4 319
AK079895.1 - 4 254
AK088702.1 - 4 316
AK152135.1 - 4 272
AK208584.1 - 2 111
BB648932.1 - 4 319
BB842333.1 - 4 293
BC023906.1 - 4 309 5321969
BC029075.1 - 4 308 4527901
BC050910.1 - 3 240 6417527

View Full Product Details