Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153563.2 | NP_705791.2 | 6, 7 | 688 | |
GenBank mRNA | AK032065.1 | - | 6, 7 | 670 | |
AK035833.1 | - | 6, 7 | 673 | ||
AK122553.1 | - | 6, 7 | 652 | ||
AK148011.1 | - | 6, 7 | 673 | ||
AK169644.1 | - | 6, 7 | 620 | ||
AK178114.1 | - | 1, 2 | 68 | ||
BC023952.1 | - | 6, 7 | 664 | 5320305 |