Gene Transcripts

Gene Symbol
Fam13a
Entrez Gene ID
58909
Gene Name
family with sequence similarity 13, member A
Gene Aliases
AI462012, D430015B01Rik, FAM13A1, Precm1
Chromosome Location
Chr. 6: 58933536 - 59024502 on Build GRCm38
UniGene ID
Mm.28813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153574.2 NP_705802.1 4 414
GenBank mRNA AJ510136.1 - 4 400
AK052421.1 - 4 355
AK054311.1 - 4 401
BC005554.1 - 4 319
BC054053.1 - 4 388
BC060978.1 - 4 351
BC080861.1 - 4 414 30357292

View Full Product Details