Gene Transcripts

Gene Symbol
Dpyd
Entrez Gene ID
99586
Gene Name
dihydropyrimidine dehydrogenase
Gene Aliases
AI315208, DPD, E330028L06Rik
Chromosome Location
Chr. 3: 118562109 - 119432918 on Build GRCm38
UniGene ID
Mm.27907
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170778.2 NP_740748.1 5 521
GenBank mRNA AK145094.1 - 5 569
AK149426.1 - 5 523
BC039699.1 - 5 522 4235971

View Full Product Details