Gene Transcripts

Gene Symbol
Fuk
Entrez Gene ID
234730
Gene Name
fucokinase
Gene Aliases
1110046B12Rik, AI303278
Chromosome Location
Chr. 8: 110882456 - 110902488 on Build GRCm38
UniGene ID
Mm.102591
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172283.3 NP_758487.2 14 1452
GenBank mRNA AJ297482.4 - 14 1423
AJ534942.1 - 14 1423
AK049153.1 - 3 281
AK131165.1 - 6 1230
AK161763.1 - 14 1554
AK164340.1 - 14 1519
AY223865.1 - 14 1423
BC037698.1 - 13 1401 4190449

View Full Product Details