Gene Transcripts

Gene Symbol
Tbc1d16
Entrez Gene ID
207592
Gene Name
TBC1 domain family, member 16
Gene Aliases
B930087K01, BC026530, Tcd1d16
Chromosome Location
Chr. 11: 119143043 - 119228499 on Build GRCm38
UniGene ID
Mm.205263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172443.3 NP_766031.2 2, 3 348
GenBank mRNA AK047555.1 - 2, 3 349
BC057634.1 - 2, 3 310 2866041
CJ111152.1 - 2, 3 320

View Full Product Details