Gene Transcripts

Gene Symbol
Atg14
Entrez Gene ID
100504663
Gene Name
autophagy related 14
Gene Aliases
4832427M01, Atg14L, D14Ertd114e, D14Ertd436e
Chromosome Location
Chr. 14: 47540893 - 47568434 on Build GRCm38
UniGene ID
Mm.287279
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172599.4 NP_766187.1 1, 2 257
GenBank mRNA AK029330.1 - 1, 2 238
AK029967.1 - 1, 2 238
AK054196.1 - 1, 2 258
BC090995.1 - 1, 2 237 6754513

View Full Product Details