Gene Transcripts

Gene Symbol
Agpat9
Entrez Gene ID
231510
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 9
Gene Aliases
1-AGPAT, 1-AGPAT 9, 4933407I02Rik, 4933408F15, 9, A230097K15Rik, GPAT-3, mGPAT3
Chromosome Location
Chr. 5: 100845710 - 100899102 on Build GRCm38
UniGene ID
Mm.271911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172715.3 NP_766303.1 3 471
GenBank mRNA AK030171.1 - 4 656
AK138410.1 - 2 631
AK160261.1 - 3 416
BC096769.1 - 3 404 30101305
BC138228.1 - 2 224 8861247
BC145669.1 - 2 224 40130972

View Full Product Details