Gene Transcripts

Gene Symbol
Shisa7
Entrez Gene ID
232813
Gene Name
shisa family member 7
Gene Aliases
Chromosome Location
Chr. 7: 4825551 - 4844789 on Build GRCm38
UniGene ID
Mm.44666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290291.1 NP_001277220.1 3 1046
NM_172737.4 NP_766325.3 3 1046
GenBank mRNA AK085127.1 - 2 268
AK145016.1 - 2 331
BY734785.1 - 2 331

View Full Product Details