Gene Transcripts
- Gene Symbol
- CDS1
- Entrez Gene ID
- 1040
- Gene Name
- CDP-diacylglycerol synthase 1
- Gene Aliases
- CDS
- Chromosome Location
- Chr. 4: 84582904 - 84651340 on Build GRCh38
- UniGene ID
- Hs.654899
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001263.3 |
NP_001254.2
|
2, 3 |
751 |
|
XM_005262687.2 |
XP_005262744.1
|
2, 3 |
747 |
|
XM_017007648.1 |
XP_016863137.1
|
2, 3 |
747 |
|
XM_017007649.1 |
XP_016863138.1
|
2, 3 |
747 |
|
XM_017007650.1 |
XP_016863139.1
|
2, 3 |
747 |
|
XM_017007651.1 |
XP_016863140.1
|
2, 3 |
747 |
|
GenBank mRNA |
AK314245.1 |
- |
2, 3 |
490 |
|
BC074833.2 |
- |
2, 3 |
302 |
30915545 |
BC074881.2 |
- |
2, 3 |
302 |
30915239 |
U60808.1 |
- |
2, 3 |
402 |
|
U65887.1 |
- |
2, 3 |
689 |
|
View Full Product Details