Gene Transcripts

Gene Symbol
Tstd2
Entrez Gene ID
272027
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 2
Gene Aliases
3010020C06
Chromosome Location
Chr. 4: 46114746 - 46138581 on Build GRCm38
UniGene ID
Mm.331588
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173033.3 NP_766621.3 3 369
GenBank mRNA AK028258.1 - 3 312
AK155456.1 - 3 316
AK165359.1 - 3 308
BC057893.1 - 3 304 4978941

View Full Product Details