Gene Transcripts

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
Chromosome Location
Chr. 4: 76011180 - 76112802 on Build GRCh38
UniGene ID
Hs.24976
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130016.2 NP_001123488.1 3 217
NM_001130017.2 NP_001123489.1 3 233
NM_001179.5 NP_001170.2 3 217
XM_005262997.1 XP_005263054.1 2 92
XM_005262999.1 XP_005263056.1 2 92
XM_005263003.1 XP_005263060.1 2 92
XM_005263004.1 XP_005263061.1 2 92
XM_006714220.1 XP_006714283.1 2 92
XM_011531971.1 XP_011530273.1 2 92
XM_017008206.1 XP_016863695.1 2 92
XM_017008207.1 XP_016863696.1 2 92
XM_017008208.1 XP_016863697.1 4 605
XM_017008209.1 XP_016863698.1 2 92
XM_017008210.1 XP_016863699.1 4 603
GenBank mRNA AK225860.1 - 3 213
AK295305.1 - 3 199
AK308810.1 - 3 199
AY543657.1 - 3 209
AY543658.1 - 4 243
AY543659.1 - 5 324
AY543660.1 - 3 202
BC008397.1 - 3 204 4244549
BC017913.1 - 3 204 4292261
BT007249.1 - 2 83
CR541664.1 - 2 83
DB451386.1 - 4 263
U47054.1 - 3 113

View Full Product Details