Gene Transcripts

Gene Symbol
Chrnb3
Entrez Gene ID
108043
Gene Name
cholinergic receptor, nicotinic, beta polypeptide 3
Gene Aliases
5730417K16Rik, Acrb3
Chromosome Location
Chr. 8: 27368711 - 27399730 on Build GRCm38
UniGene ID
Mm.110444
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027454.4 NP_081730.1 4 635
NM_173212.4 NP_775304.1 5 680
GenBank mRNA AF325350.1 - 4 288
AF467896.1 - 5 443
AK017571.1 - 4 565
AK030464.1 - 5 681
AK133689.1 - 5 624
AY574268.1 - 5 422
BC058193.1 - 5 443 5357762

View Full Product Details