Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163512.1 | NP_001156984.1 | 14 | 3349 | |
NM_173402.2 | NP_775578.2 | 17 | 5396 | ||
GenBank mRNA | AK014569.1 | - | 14 | 3351 | |
BC040396.1 | - | 17 | 5393 | 4461061 |