Gene Transcripts

Gene Symbol
Psapl1
Entrez Gene ID
76943
Gene Name
prosaposin-like 1
Gene Aliases
2310020A21Rik
Chromosome Location
Chr. 5: 36204021 - 36206568 on Build GRCm38
UniGene ID
Mm.44242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175249.3 NP_780458.2 1 146
GenBank mRNA AK009408.1 - 1 136
AK028455.1 - 1 111
AK078699.1 - 1 111
BC119540.2 - 1 111
BC148306.2 - 1 111
BY770175.1 - 1 147

View Full Product Details