Gene Transcripts

Gene Symbol
Fbxl22
Entrez Gene ID
74165
Gene Name
F-box and leucine-rich repeat protein 22
Gene Aliases
1110004B15Rik
Chromosome Location
Chr. 9: 66508459 - 66514593 on Build GRCm38
UniGene ID
Mm.297998
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175206.4 NP_780415.1 2 788
GenBank mRNA AK052175.1 - 2 774
AK084845.1 - 2 704
AK085980.1 - 2 731

View Full Product Details