Gene Transcripts

Gene Symbol
Ttyh3
Entrez Gene ID
78339
Gene Name
tweety homolog 3 (Drosophila)
Gene Aliases
2900029G13Rik, AI414930, TTY3, mTTY3
Chromosome Location
Chr. 5: 140620578 - 140649031 on Build GRCm38
UniGene ID
Mm.28947
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK052340.1 - 14 3652
AK173233.1 - 11 3041
AK194537.1 - 1 107
BC059083.1 - 13 3575 6827289
BC062917.1 - 14 3658 6831835

View Full Product Details