Gene Transcripts

Gene Symbol
Rspo4
Entrez Gene ID
228770
Gene Name
R-spondin family, member 4
Gene Aliases
A730099F22, A930029K19Rik
Chromosome Location
Chr. 2: 151842579 - 151874668 on Build GRCm38
UniGene ID
Mm.159868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040689.1 NP_001035779.1 1, 2 223
GenBank mRNA AK020904.1 - 1, 2 224
AK043484.1 - 1, 2 224
BC048707.1 - 1, 2 197 6511472
BC116367.1 - 1, 2 186

View Full Product Details