Gene Transcripts

Gene Symbol
Gphb5
Entrez Gene ID
217674
Gene Name
glycoprotein hormone beta 5
Gene Aliases
OGH, Zlut1
Chromosome Location
Chr. 12: 75411720 - 75416781 on Build GRCm38
UniGene ID
Mm.252250
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175644.3 NP_783575.2 3 568
GenBank mRNA AK078727.1 - 3 570
BC111115.1 - 2 409
BC139462.1 - 3 508 8862484
BC139463.1 - 3 508 8862485

View Full Product Details