Gene Transcripts

Gene Symbol
Dnajc22
Entrez Gene ID
72778
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 22
Gene Aliases
2810451A06Rik, AI506245
Chromosome Location
Chr. 15: 99099134 - 99104737 on Build GRCm38
UniGene ID
Mm.30544
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176835.2 NP_789805.1 2 1051
GenBank mRNA AK013323.1 - 2 1074
AK050294.1 - 2 1030
BC039633.1 - 2 1051 5136672

View Full Product Details