Gene Transcripts

Gene Symbol
Ust
Entrez Gene ID
338362
Gene Name
uronyl-2-sulfotransferase
Gene Aliases
D930010O20Rik, UA2OST
Chromosome Location
Chr. 10: 8204753 - 8518825 on Build GRCm38
UniGene ID
Mm.41163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177387.3 NP_796361.2 3 749
GenBank mRNA AK086181.1 - 3 696
AK147850.1 - 3 750
AK162950.1 - 3 322
BC138155.1 - 3 489 8861174

View Full Product Details