Gene Transcripts

Gene Symbol
Igfn1
Entrez Gene ID
226438
Gene Name
immunoglobulin-like and fibronectin type III domain containing 1
Gene Aliases
9830123M21Rik
Chromosome Location
Chr. 1: 135953205 - 136040842 on Build GRCm38
UniGene ID
Mm.119675
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177642.3 NP_808310.2 27 8904
GenBank mRNA AK036509.1 - 8 1679
EU429481.1 - 13 5776
EU429482.1 - 12 2629
GU290042.1 - 27 8904

View Full Product Details