Gene Transcripts

Gene Symbol
Kctd19
Entrez Gene ID
279499
Gene Name
potassium channel tetramerisation domain containing 19
Gene Aliases
4922504H04Rik
Chromosome Location
Chr. 8: 105382350 - 105413502 on Build GRCm38
UniGene ID
Mm.67628
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301173.1 NP_001288102.1 4 564
NM_177791.3 NP_808459.1 4 564
GenBank mRNA AK076644.1 - 4 556
BC092530.1 - 4 564 6744085
BC125431.1 - 4 556 40129946
BC125433.1 - 4 556 40129948

View Full Product Details