Gene Transcripts

Gene Symbol
Urgcp
Entrez Gene ID
72046
Gene Name
upregulator of cell proliferation
Gene Aliases
2010005J08Rik, 9130001I21Rik, AW060359, Urg4, mKIAA1507
Chromosome Location
Chr. 11: 5713417 - 5762436 on Build GRCm38
UniGene ID
Mm.271657
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077661.1 NP_001071129.1 6 826
NM_178623.3 NP_848738.2 6 410
GenBank mRNA AK033577.1 - 6 384
AK035580.1 - 6 822
AK041953.1 - 7 1075
AK129378.1 - 5 468
AK166495.1 - 5 310
AK190812.1 - 3 199
AK211902.1 - 2 177
BC066994.1 - 6 707 5696000

View Full Product Details