Gene Transcripts

Gene Symbol
ADH1C
Entrez Gene ID
126
Gene Name
alcohol dehydrogenase 1C (class I), gamma polypeptide
Gene Aliases
ADH3
Chromosome Location
Chr. 4: 99336492 - 99353045 on Build GRCh38
UniGene ID
Hs.654537
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000669.4 NP_000660.1 7, 8 1340
NR_133005.1 7, 8 1296
XM_011531588.2 XP_011529890.1 6, 7 1130
XM_011531589.1 XP_011529891.1 8, 9 1460
GenBank mRNA BC062476.1 - 7, 8 1036
BC066227.1 - 7, 8 998 7002195
BC066228.1 - 7, 8 998 7002197
BC067419.1 - 7, 8 998 7002191
BC067420.1 - 7, 8 998 7002192
BC067421.1 - 7, 8 998 7002193
BC067422.1 - 7, 8 998 7002194
BC074771.2 - 7, 8 998 30915457
BC074786.2 - 7, 8 997 30915344
CD014078.1 - 7, 8 997
M12272.1 - 7, 8 1050
X04299.1 - 7, 8 1042
X04350.1 - 5, 6 812

View Full Product Details