Gene Transcripts

Gene Symbol
AMBN
Entrez Gene ID
258
Gene Name
ameloblastin
Gene Aliases
AI1F
Chromosome Location
Chr. 4: 70592258 - 70607288 on Build GRCh38
UniGene ID
Hs.272396
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016519.5 NP_057603.1 13 1380
GenBank mRNA AF209780.1 - 13 1362
AF219994.2 - 13 1344
AF263464.1 - 13 1354
AJ537437.1 - 13 1354
AK314918.1 - 13 1315
BC106931.1 - 13 1315 40033703
BC106932.1 - 13 1312 40033705

View Full Product Details