Gene Transcripts
- Gene Symbol
- FAS
- Entrez Gene ID
- 355
- Gene Name
- Fas cell surface death receptor
- Gene Aliases
- ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
- Chromosome Location
- Chr. 10: 88968429 - 89017061 on Build GRCh38
- UniGene ID
- Hs.244139
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000043.5 |
NP_000034.1
|
1, 2 |
358 |
|
NM_001320619.1 |
NP_001307548.1
|
1, 2 |
358 |
|
NM_152871.3 |
NP_690610.1
|
1, 2 |
358 |
|
NM_152872.3 |
NP_690611.1
|
1, 2 |
358 |
|
NR_028033.3 |
|
1, 2 |
358 |
|
NR_028034.3 |
|
1, 2 |
358 |
|
NR_028035.3 |
|
1, 2 |
358 |
|
NR_028036.3 |
|
1, 2 |
358 |
|
NR_135313.1 |
|
1, 2 |
358 |
|
XM_006717819.3 |
XP_006717882.1
|
2, 3 |
5040 |
|
XM_011539764.2 |
XP_011538066.1
|
2, 3 |
429 |
|
XM_011539765.2 |
XP_011538067.1
|
2, 3 |
217 |
|
XM_011539766.2 |
XP_011538068.1
|
2, 3 |
169 |
|
XM_011539767.2 |
XP_011538069.1
|
1, 2 |
92 |
|
GenBank mRNA |
AB209361.1 |
- |
2, 3 |
3675 |
|
AK290978.1 |
- |
1, 2 |
286 |
|
AY495076.2 |
- |
1, 2 |
234 |
|
BC012479.1 |
- |
1, 2 |
350 |
4514272 |
CR450307.1 |
- |
1, 2 |
40 |
|
DA699562.1 |
- |
1, 2 |
358 |
|
EU481974.2 |
- |
1, 2 |
234 |
|
FJ200481.1 |
- |
1, 2 |
40 |
|
FJ373045.1 |
- |
1, 2 |
40 |
|
FJ373046.1 |
- |
1, 2 |
40 |
|
FM246458.1 |
- |
1, 2 |
40 |
|
FM246459.1 |
- |
1, 2 |
40 |
|
M67454.1 |
- |
1, 2 |
234 |
|
X63717.1 |
- |
1, 2 |
260 |
|
X83490.1 |
- |
1, 2 |
65 |
|
X83491.1 |
- |
1, 2 |
65 |
|
X83492.1 |
- |
1, 2 |
65 |
|
X83493.1 |
- |
1, 2 |
65 |
|
X89101.1 |
- |
1, 2 |
40 |
|
Z47993.1 |
- |
1, 2 |
40 |
|
Z47994.1 |
- |
1, 2 |
40 |
|
Z47995.1 |
- |
1, 2 |
40 |
|
Z66556.1 |
- |
1, 2 |
40 |
|
Z70519.1 |
- |
1, 2 |
40 |
|
Z70520.1 |
- |
1, 2 |
40 |
|
View Full Product Details