Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006713314.3 | XP_006713377.1 | 5 | 1858 | |
XM_011534041.2 | XP_011532343.1 | 5 | 1695 | ||
XM_017007088.1 | XP_016862577.1 | 6 | 1971 | ||
GenBank mRNA | AK294301.1 | - | 6 | 1710 | |
AK297033.1 | - | 4 | 1570 | ||
AK301838.1 | - | 4 | 1770 | ||
AK313252.1 | - | 4 | 1561 | ||
AL573841.3 | - | 1 | 478 | ||
BC012099.1 | - | 4 | 1504 | 4559618 | |
BM565546.1 | - | 1 | 503 | ||
U03274.1 | - | 4 | 1507 |