Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001226.3 | NP_001217.2 | 7 | 879 | |
NM_032992.2 | NP_116787.1 | 4 | 612 | ||
NR_133012.1 | 6 | 802 | |||
GenBank mRNA | BC000305.2 | - | 7 | 827 | |
BC004460.1 | - | 7 | 827 | 2820468 | |
BM837197.1 | - | 4 | 558 | ||
U20536.1 | - | 7 | 879 | ||
U20537.1 | - | 4 | 534 |