Gene Transcripts

Gene Symbol
CDC5L
Entrez Gene ID
988
Gene Name
cell division cycle 5 like
Gene Aliases
CDC5, CDC5-LIKE, CEF1, PCDC5RP, dJ319D22.1
Chromosome Location
Chr. 6: 44387514 - 44450424 on Build GRCh38
UniGene ID
Hs.485471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253.3 NP_001244.1 8 1290
GenBank mRNA AB007892.2 - 8 1062
AK128737.1 - 8 1098
AK299738.1 - 8 1017
BC001568.1 - 8 1079 3452352
U86753.1 - 8 980

View Full Product Details