Gene Transcripts

Gene Symbol
SEPT7
Entrez Gene ID
989
Gene Name
septin 7
Gene Aliases
CDC10, CDC3, NBLA02942, SEPT7A
Chromosome Location
Chr. 7: 35800986 - 35915763 on Build GRCh38
UniGene ID
Hs.191346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011553.3 NP_001011553.2 2, 3 387
NM_001788.5 NP_001779.3 3, 4 390
XM_006715806.3 XP_006715869.1 3, 4 338
XM_011515656.2 XP_011513958.1 3, 4 338
XM_011515661.2 XP_011513963.1 3, 4 338
XM_017012855.1 XP_016868344.1 3, 4 322
XM_017012856.1 XP_016868345.1 3, 4 330
XM_017012857.1 XP_016868346.1 3, 4 327
XM_017012862.1 XP_016868351.1 2, 3 153
XM_017012863.1 XP_016868352.1 3, 4 696
GenBank mRNA AB073389.1 - 3, 4 157
AB209677.1 - 3, 4 125
AK223225.1 - 3, 4 157
AK223241.1 - 3, 4 157
AK290545.1 - 2, 3 228
AK298453.1 - 3, 4 224
BC025987.2 - 2, 3 154 4823810
BC067264.1 - 2, 3 133 6503495
BC093640.1 - 3, 4 156 7939485
BC093642.1 - 3, 4 156 7939487
BC107718.1 - 2, 3 335
BI669760.1 - 3, 4 368
BP280069.1 - 3, 4 361
BQ229116.1 - 2, 3 318
BQ232357.1 - 3, 4 132
BX571755.1 - 2, 3 155
DC414394.1 - 2, 3 390
DQ232879.1 - 2, 3 138
S72008.1 - 3, 4 157

View Full Product Details